missing translation for 'onlineSavingsMsg'
Learn More

Neomycin sulfate, MedChemExpress

Product Code. 16478498
Encompass_Preferred
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 16478498

Brand: Medchem Express HYB0470

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neomycin sulfate, an aminoglycoside antibiotic, exerts antibacterial activity through irreversible binding of the nuclear 30S ribosomal subunit, thereby blocking bacterial protein synthesis. Neomycin sulfate is a known phospholipase C (PLC) inhibitor. Neomycin sulfate potently inhibits both nuclear translocation of angiogenin and angiogenin-induced cell proliferation and angiogenesis.

Neomycin sulfate, an aminoglycoside antibiotic, exerts antibacterial activity through irreversible binding of the nuclear 30S ribosomal subunit, thereby blocking bacterial protein synthesis. Neomycin sulfate is a known phospholipase C (PLC) inhibitor. Neomycin sulfate potently inhibits both nuclear translocation of angiogenin and angiogenin-induced cell proliferation and angiogenesis.

Specifications

Chemical Name or Material Neomycin sulfate
CAS 1405-10-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H52N6O25S3
Quantity 10 g
Solubility Information H2O : 250 mg/mL (275.06 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](O)[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@H]2O[C@@H]3O[C@H](CO)[C@@H](O[C@H]4O[C@@H](CN)[C@@H](O)[C@H](O)[C@H]4N)[C@H]3O)O[C@@H]1CN.[3H2SO4]
Molecular Weight (g/mol) 908.88
Formula Weight 908.88
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.