missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neomycin sulfate, MedChemExpress
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
Description
Neomycin sulfate, an aminoglycoside antibiotic, exerts antibacterial activity through irreversible binding of the nuclear 30S ribosomal subunit, thereby blocking bacterial protein synthesis. Neomycin sulfate is a known phospholipase C (PLC) inhibitor. Neomycin sulfate potently inhibits both nuclear translocation of angiogenin and angiogenin-induced cell proliferation and angiogenesis.
Specifications
Specifications
| Chemical Name or Material | Neomycin sulfate |
| CAS | 1405-10-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H52N6O25S3 |
| Quantity | 25 g |
| Solubility Information | H2O : 250 mg/mL (275.06 mM; Need ultrasonic) |
| SMILES | O[C@H]1[C@H](O)[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@H]2O[C@@H]3O[C@H](CO)[C@@H](O[C@H]4O[C@@H](CN)[C@@H](O)[C@H](O)[C@H]4N)[C@H]3O)O[C@@H]1CN.[3H2SO4] |
| Molecular Weight (g/mol) | 908.88 |
| Formula Weight | 908.88 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction