missing translation for 'onlineSavingsMsg'
Learn More

Melford Neomycin Sulfate

Product Code. 30018564
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018564 25 g 25g
30018563 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018564 Supplier Melford Supplier No. N2004025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neomycin Sulfate is an antibiotic that is derived from the bacterium Streptomyces fradiae and belongs to the aminoglycoside class of antibiotics.

Neomycin Sulfate is an antibiotic that is derived from the bacterium Streptomyces fradiae and belongs to the aminoglycoside class of antibiotics. It exhibits broad-spectrum activity against Gram-negative and some Gram-positive bacteria.

In molecular biology, Neomycin Sulfate is commonly used as a selective agent in genetic engineering experiments and cell culture applications. It is often included in growth media or buffers to prevent the growth of bacteria and ensure the purity of cell lines.

Neomycin Sulfate functions by inhibiting protein synthesis in bacteria, leading to cell death. Neomycin Sulfate is also employed in microbiology research. It can be used to selectively isolate and identify bacteria or as a control for susceptibility testing in antimicrobial resistance studies.

Specifications

Chemical Name or Material Neomycin Sulfate
CAS 1405-10-3
Quantity 25 g
Molecular Formula C23H46N6O13-3H2OSO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.