missing translation for 'onlineSavingsMsg'
Learn More

Neohesperidin-dihydrochalcone, Mikromol™

Product Code. 17919964
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17919964 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17919964 Supplier Mikromol Supplier No. MM3398.00
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Neohesperidin-dihydrochalcone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Neohesperidin Dihydrochalcone
CAS 20702-77-6
Analyte or Component Names Neohesperidin Dihydrochalcone
Shipping Condition Room Temperature
Molecular Formula C28 H36 O15
Certifications/Compliance ISO 17034
Synonym Neohesperidin-dihydrochalcone, Citrosa, NHDC, Neohesperidin DC, Neohesperidin DHC, Neohesperidin dihydrochalcone, Neohesperidine dihydrochalcone, 1-[4-[[2-O-(6-Deoxy-alpha-l-mannopyranosyl)-beta-d-glucopyranosyl]oxy]-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)propan-1-one, 1-Propanone, 1-[4-[[2-O-(6-deoxy-alpha-L-mannopyranosyl)-beta-D-glucopyranosyl]oxy]-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)-, Glucopyranoside, 3,5-dihydroxy-4-(3-hydroxy-4-methoxyhydrocinnamoyl)phenyl 2-O-(6-deoxy-alpha-L-mannopyranosyl)-, beta-D- (8CI), Glucopyranoside, 3,5-dihydroxy-4-(3-hydroxy-4-methoxyhydrocinnamoyl)phenyl 2-O-alpha-L-rhamnopyranosyl- (7CI), 1-[4-[[2-O-(6-Deoxy-alpha-L-mannopyranosyl)-beta-D-glucopyranosyl]oxy]-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)-1-propanone
InChI Formula InChI=1S/C28H36O15/c1-11-21(34)23(36)25(38)27(40-11)43-26-24(37)22(35)19(10-29)42-28(26)41-13-8-16(32)20(17(33)9-13)14(30)5-3-12-4-6-18(39-2)15(31)7-12/h4,6-9,11,19,21-29,31-38H,3,5,10H2,1-2H3/t11-,19+,21-,22+,23+,24-,25+,26+,27-,28+/m0/s1
SMILES COc1ccc(CCC(=O)c2c(O)cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)cc2O)cc1O
IUPAC Name 1-[4-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)propan-1-one
Molecular Weight (g/mol) 612.58
Quantity 250 mg
Formula Weight 612.2054 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.