missing translation for 'onlineSavingsMsg'
Learn More

ICU Medical Neoheel™ Safety Lancets

Product Code. 17211822
Encompass
Change view
Click to view available options
Quantity:
50/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17211822 50/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17211822 Supplier ICU Medical Supplier No. 1052N

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neoheel™ Safety Lancets for collecting quality blood samples from newborns

  • Used to collect quality blood samples from infants for metabolic screening as well as glucose and other tests
  • Automatic retraction is designed to avoid needlestick injuries and cross-contamination exposure
  • The safety lancet produces less bruising and requires less punctures to obtain sufficient volumes of blood
  • Arc shaped design reduces the number of heel puncture sites, time taken to complete the test and time the baby cries
  • Integral safety tab prevents premature activation while avoiding additional loose parts

Specifications

Color Yellow
Disposable Single-use
For Use With (Application) Newborn Blood Collection
Quantity 50/Box
Product Type Safety Lancet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.