missing translation for 'onlineSavingsMsg'
Learn More

Neogenin Antibody (RM0124-3G55), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18431534 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431534 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431534 Supplier Novus Biologicals Supplier No. NBP122501MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Neogenin Monoclonal antibody specifically detects Neogenin in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neogenin
Applications Western Blot
Classification Monoclonal
Clone RM0124-3G55
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGNDKFZp547A066
Host Species Rat
Immunogen Mouse recombinant protein of Neogenin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4756
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.