missing translation for 'onlineSavingsMsg'
Learn More

Neodymium standard solution, for AAS, 1mg/mL Nd in 2% HNO3, Thermo Scientific™

Product Code. 10616972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10616972 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10616972 Supplier Thermo Scientific Acros Supplier No. 196141000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-00-8 , 7697-37-2 , 7732-18-5
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

Packaging Plastic bottle
CAS Min % 1.9, 96.9
CAS Max % 2.1, 97.1
Physical Form Solution
Concentration 995 to 1005 μg/mL of Nd in 2% HNO3 (20°C)
Quantity 100 mL
Formula Weight 144.24
Grade AAS
Chemical Name or Material Neodymium standard solution, 1 mg/ml Nd in 2% HNO3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.