missing translation for 'onlineSavingsMsg'
Learn More

Neodymium rod, 6.35mm (0.25in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals

Catalog Number 11315118
Click to view available options
Quantity:
10 mm
50 mm
100 mm
This item is not returnable. View return policy
7440-00-8
Nd
144.24
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
23934
CHEBI:33372
neodymium
[Nd]
This item is not returnable. View return policy
7440-00-8
Nd
144.24
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
23934
CHEBI:33372
neodymium
[Nd]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mylar under argon
TRUSTED_SUSTAINABILITY
Quantity 10 mm
Percent Purity 99.5%
Odor Odorless
Packaging Mylar under Argon
Weight ≈2.2g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Neodymium rod, ≈6.35mm (0.25 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'