missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) sulfate octahydrate, 99.9% (REO)

Product Code. 11383768
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11383768

Brand: Thermo Scientific Alfa Aesar 040253.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13477-91-3 | Nd2(SO4)3·8H2O

It is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-91-3
Molecular Formula Nd2(SO4)3·8H2O
MDL Number MFCD00149803

Specifications

Melting Point 1176°C
Color Pink
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.9%
Solubility Information Soluble in water
Formula Weight 720.78 (576.66 Anhydrous)
Odor Odorless
Appearance Pink crystals
Density 2.85
Chemical Name or Material Neodymium(III) sulfate octahydrate, Rare Earth Oxides Content
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.