missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals

Catalog Number 11363768
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1313-97-9
Nd2O3
336.48
MFCD00011134
PLDDOISOJJCEMH-UHFFFAOYSA-N
dineodymium(3+) trioxidandiide
[O--].[O--].[O--].[Nd+3].[Nd+3]
This item is not returnable. View return policy
1313-97-9
Nd2O3
336.48
MFCD00011134
PLDDOISOJJCEMH-UHFFFAOYSA-N
dineodymium(3+) trioxidandiide
[O--].[O--].[O--].[Nd+3].[Nd+3]

ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst.Neodymium(III) oxide is an inorganic compound containing inner transition metal neodymium from lanthanides series in +3 oxidation state and oxide in 2:3 ratio. It adopts a hexagonal crystal structure. It is also known as neodymium oxide, neodymiumtrioxide, neodymium(3+)oxide, neodymium sesquioxide and dineodymiumtrioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst
TRUSTED_SUSTAINABILITY
Melting Point 2272°C
Physical Form Powder
Quantity 25 g
Merck Index 14,6450
Solubility Information Insoluble in water; Soluble in acids.
Formula Weight 336.48
Percent Purity 99%
Grade Reacton™
Density 7.24 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'