missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11346308
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
1313-97-9
Nd2O3
336.48
MFCD00011134
PLDDOISOJJCEMH-UHFFFAOYSA-N
dineodymium(3+) trioxidandiide
[O--].[O--].[O--].[Nd+3].[Nd+3]
This item is not returnable. View return policy
1313-97-9
Nd2O3
336.48
MFCD00011134
PLDDOISOJJCEMH-UHFFFAOYSA-N
dineodymium(3+) trioxidandiide
[O--].[O--].[O--].[Nd+3].[Nd+3]

Neodymium(III) oxide is used as a catalyst in polymer chemistry. Glasses doped with neodymium(III) oxide are dichroic in nature and is used in welding goggles and other colored glasses. It finds use in solid-state lasers, carbon arc-light electrodes and enamels. It serves as the starting material for the synthesis of other neodymium(III) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2272°C
Color Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 336.48
Appearance Light blue powder
Density 7.24 g/cm3 at 20°C
Chemical Name or Material Neodymium(III) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'