missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) nitrate hexahydrate, 99.9% (REO)

Product Code. p-7049104
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11327647

Brand: Thermo Scientific Alfa Aesar 012912.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16454-60-7 | H12N3NdO15 | 438.344 g/mol

Neodymium(III) nitrate hexahydrate is used to prepare nanoparticles of neodymium oxybromide and neodymium oxide. It is also used in coloring glasses and for doping some glass lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16454-60-7
Molecular Formula H12N3NdO15
Molecular Weight (g/mol) 438.344
MDL Number MFCD00149799
InChI Key VQVDTKCSDUNYBO-UHFFFAOYSA-N
Synonym neodymium nitrate hexahydrate, neodymium trinitrate hexahydrate, neodymium nitrate monohydrate, neodymium iii nitrate, hexahydrate 1:3:6, nitric acid, neodymium 3+ salt, hexahydrate, ksc491s0r, neodymiumnitratehexahydrate, neodymium nitrate 6-hydrate, neodymium 3+ trinitrate hexahydrate, neodymium iii nitrate hexahydrate
PubChem CID 204494
IUPAC Name neodymium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3]

Specifications

Melting Point ∼40°C
pH 3.0 to 4.0
Physical Form Crystalline
Quantity 50 g
UN Number UN1477
Merck Index 14,6450
Solubility Information Soluble in water and alcohol.
Formula Weight 438.34 (330.25 Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Neodymium(III) nitrate hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Neodymium(III) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Neodymium(III) nitrate, hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.