missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) isopropoxide, 99% (REO), 2.5% w/v in toluene/isopropanol

Product Code. 11328008
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328008 100 mL 100mL
11338008 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328008 Supplier Thermo Scientific Alfa Aesar Supplier No. 036573.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19236-15-8 | C9H24NdO3 | 324.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19236-15-8
Molecular Formula C9H24NdO3
Molecular Weight (g/mol) 324.53
MDL Number MFCD00058819,MFCD00058819
InChI Key WOCLHTSADRFWMF-UHFFFAOYSA-N
Synonym neodymium i-propoxide, neodymium iii isopropoxide
PubChem CID 25199945
SMILES [Nd].CC(C)O.CC(C)O.CC(C)O

Specifications

Color Blue
Linear Formula Nd[OCH(CH3)2]3
Quantity 100 mL
UN Number UN1993
Sensitivity Moisture sensitive
IUPAC Name neodymium;propan-2-ol
Formula Weight 321.3
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 2.5% w/v in toluene/isopropanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Neodymium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.