missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) fluoride, anhydrous, 99.9% (REO)

Product Code. 11326308
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326308 100 g 100g
11336308 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326308 Supplier Thermo Scientific Alfa Aesar Supplier No. 011246.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13709-42-7 | F3Nd | 201.24 g/mol

Neodymium(III) fluoride, is used extensively in the preparation and study of fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-42-7
Molecular Formula F3Nd
Molecular Weight (g/mol) 201.24
MDL Number MFCD00011135
InChI Key XRADHEAKQRNYQQ-UHFFFAOYSA-K
IUPAC Name neodymium(3+) trifluoride
SMILES [F-].[F-].[F-].[Nd+3]

Specifications

Melting Point 1410°C
Color Purple
Physical Form Crystalline Lump
Quantity 100 g
Linear Formula NdF3
Solubility Information Soluble in aqueous solutions.
Formula Weight 201.24
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.5 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.