missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) chloride, anhydrous, 99.9% (REO)

Product Code. 11363457
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363457 100 g 100g
11373457 5 g 5g
11353457 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363457 Supplier Thermo Scientific Alfa Aesar Supplier No. 018680.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10024-93-8

Neodymium(III) chloride is used as a precursor for the production of neodymium metal. It is used as a catalyst and accelerates polymerization of various dienes such as polybutylene, polybutadiene, and polyisoprene. It has luminescence property and is widely used as fluorescent label in organic molecules, thereby helping in easy tracking of the compound using fluorescence microscope during various physical and chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10024-93-8
MDL Number MFCD00011132

Specifications

Melting Point 784°C
Physical Form Powder
Quantity 100 g
Merck Index 14,6450
Solubility Information Soluble in water and ethanol.
Formula Weight 250.6
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 4.134 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.