missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals

£120.00 - £376.00

Chemical Identifiers

CAS 38245-38-4
Molecular Formula C3Nd2O9
Molecular Weight (g/mol) 468.51
MDL Number MFCD00150426
InChI Key UTWHRPIUNFLOBE-UHFFFAOYSA-H
Synonym neodymium carbonate 2:3 hydrate, neodymium carbonate hydrate, carbonic acid, neodymium 3+ salt 3:2 , hydrate, neodymium iii carbonate hydrate, neodymium carbonate hydrate 2:3:1, 3co3.2nd.h2o, neodymium 3+ tricarbonate hydrate, dineodymium 3+ ion hydrate trico3, neodymium carbonate, dineodymium 3+ hydrate tricarbonate
PubChem CID 217202
IUPAC Name neodymium(3+);tricarbonate;hydrate
SMILES [Nd+3].[Nd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377118
View Documents Promotion Details
Thermo Scientific Alfa Aesar
015301.18
50 g
£120.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11367118
View Documents Promotion Details
Thermo Scientific Alfa Aesar
015301.30
250 g
£376.00
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Neodymium Carbonate mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38245-38-4
468.51
UTWHRPIUNFLOBE-UHFFFAOYSA-H
217202
[Nd+3].[Nd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
C3Nd2O9
MFCD00150426
neodymium carbonate 2:3 hydrate, neodymium carbonate hydrate, carbonic acid, neodymium 3+ salt 3:2 , hydrate, neodymium iii carbonate hydrate, neodymium carbonate hydrate 2:3:1, 3co3.2nd.h2o, neodymium 3+ tricarbonate hydrate, dineodymium 3+ ion hydrate trico3, neodymium carbonate, dineodymium 3+ hydrate tricarbonate
neodymium(3+);tricarbonate;hydrate

Specifications

38245-38-4
C3Nd2O9
neodymium carbonate 2:3 hydrate, neodymium carbonate hydrate, carbonic acid, neodymium 3+ salt 3:2 , hydrate, neodymium iii carbonate hydrate, neodymium carbonate hydrate 2:3:1, 3co3.2nd.h2o, neodymium 3+ tricarbonate hydrate, dineodymium 3+ ion hydrate trico3, neodymium carbonate, dineodymium 3+ hydrate tricarbonate
UTWHRPIUNFLOBE-UHFFFAOYSA-H
neodymium(3+);tricarbonate;hydrate
217202
99.9%
Neodymium(III) carbonate hydrate
Powder
MFCD00150426
Insoluble in water,moderately soluble in strong mineral acids.
[Nd+3].[Nd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
468.51
468.51 (Anhydrous)
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 227-579-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only