missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)

Product Code. 11302568
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302568 25 g 25g
11312568 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302568 Supplier Thermo Scientific Alfa Aesar Supplier No. 014606.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29843-90-1 | Br3Nd | 383.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29843-90-1
Molecular Formula Br3Nd
Molecular Weight (g/mol) 383.95
MDL Number MFCD00748137
InChI Key LBWLQVSRPJHLEY-UHFFFAOYSA-K
IUPAC Name neodymium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Nd+3]

Specifications

Physical Form Crystalline
Quantity 25 g
Formula Weight 383.97 (Anhydrous)
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Neodymium(III) bromide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.