missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) acetate hydrate, 99.9% (REO)

Product Code. 11455370
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455370 250 g 250g
11445370 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455370 Supplier Thermo Scientific Alfa Aesar Supplier No. 044587.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 334869-71-5 | C6H9NdO6 | 321.37 g/mol

Neodymium Acetate, mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334869-71-5
Molecular Formula C6H9NdO6
Molecular Weight (g/mol) 321.37
MDL Number MFCD00013040
InChI Key KETUDCKOKOGBJB-UHFFFAOYSA-K
IUPAC Name neodymium(3+) triacetate
SMILES [Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline
Quantity 250 g
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
Formula Weight 321.38 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Neodymium (III) acetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.