missing translation for 'onlineSavingsMsg'
Learn More

Neodymium Boron Iron, REacton™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 013642.18

Code : NEW

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 11396888

  • £64.10 / 50g
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Neodymium Boron Iron applied to manufacture head actuators for computer hard disks, magnetic resonance imaging (MRI), magnetic guitar pickups, loudspeakers and headphones, mechanical e-cigarette firing switches and magnetic bearings and couplings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Neodymium Boron Iron
Gray-Silver
50 g
MFCD02091729
Insoluble in water.
Silver gray broken lump
Magnet Alloy Additive
Broken Lump
Nd:B:Fe; 15:8:77 atomic%
Magnet Alloy Additive
Odorless
7.5g/cm3
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only