missing translation for 'onlineSavingsMsg'
Learn More

Neodymium @ 5000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17941576
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17941576 200 g 200g
17990976 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17941576 Supplier VHG Labs Supplier No. VHGOND5000200G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Tilføj til indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Neodymium @ 5000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Neodymium
CAS 7440-00-8
Methods ICP/ICPMS
Analyte or Component Names Neodymium
Shipping Condition Room Temperature
Molecular Formula Nd
Concentration 1000 μg/g
Certifications/Compliance ISO 17034
Synonym Nd
InChI Formula InChI=1S/Nd
SMILES [Nd]
IUPAC Name neodymium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 144.24
Quantity 200 g
Formula Weight 143.91 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.