missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dr.Weigert™ Neodisher MediClean Forte
Click to view available options
Volume (Metric):
20 L
Unit Size:
20 L
Description
- Enzymatic detergent, liquid concentrate
- Neodisher MediClean Forte removes Residues of dried and denatured blood are reliably removed
- Contains alkalinity donors, enzymes and surfactants
- Excellent material compatibility, universally applicable
- Can be used in manual, immersion, waher disinfection and ultra sonic bath
- Excellent material compatibility
- A redeposition of protein residues is prevented
- Dosage between 2-10 mL/L
Specifications
Specifications
| Form | Liquid |
| pH | 10.4 to 10.8 |
| Sterility | Non-sterile |
| Composition | <5 % non-ionic and anionic surfactants also: enzymes |
| Concentration | 2 to 10 mL/L |
| Container Type | Canister |
| Content And Storage | Between 0°C to 25°C. Usable for 2 years when stored as recommended. |
| Density | 1.1 g/cm3 (20°C) |
| Description | 390 x 290 x 240 mm |
| Filtered | Filtered |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction