missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher 30

Product Code. 10371371
Change view
Click to view available options
Volume (Metric):
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10371371 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10371371 Supplier Dr.Weigert™ Supplier No. 320166
  • ONLINE EXCLUSIVE PRICE
    £205.00
    £176.85 / 25kg
    Save £28.15
    13% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher 30 - Universal detergent powder for automated dishwashing. Contains active chlorine for superior cleaning results, even with tea and coffee residues.

  • Alkaline detergent, Powder
  • neodisher 30 is used for automated cleaning of tableware, cutlery working utensils and glasses in all food processing companies such as professional kitchens, butcher’s shops and bakeries
  • Removes especially protein, coffee and tea residues
  • High level of material protection
  • Dosage of 2-4 g/L
  • Usable with all degrees of water hardness

Specifications

Product Type Neodisher 30
Form Powder
pH 11.5 to 11.7
Sterility Non-sterile
Composition <5 % bleaching agent based on chlorine > 30 % phosphates
Concentration 2 to 4 g/L
Density 1,050 g/L
Description 600 x 300 x 150 mm
Volume (Metric) 25 kg
compliance-icons compliance-icons
Product Identifier
  • neodisher 30
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: disodium metasilicate pentahydrate, sodium carbonate, troclosene sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.