missing translation for 'onlineSavingsMsg'
Learn More

Neocuproine hemihydrate, 99+%

Product Code. 10414025 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10414025 25 g Glass bottle 25g
10776161 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10414025 Supplier Thermo Scientific Acros Supplier No. 153320250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34302-69-7 | C28H26N4O | 434.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34302-69-7
Molecular Formula C28H26N4O
Molecular Weight (g/mol) 434.54
MDL Number MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843
InChI Key IEBXFSLFDFHSRD-UHFFFAOYSA-N
Synonym unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate
PubChem CID 67652146
SMILES O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 158.0°C to 163.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Total base)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 6534
Solubility Information Solubility in water: slightly soluble. Other solubilities: 1500g/L methanol (20°C)
IUPAC Name 2,9-dimethyl-1,10-phenanthroline;hydrate
Formula Weight 217.27
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Neocuproine hemihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Neocuproine hemihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.