missing translation for 'onlineSavingsMsg'
Learn More

Neobavaisoflavone, MedChemExpress

Product Code. 16441488
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441488 5 mg 5mg
16419078 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441488 Supplier Medchem Express Supplier No. HYN0720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neobavaisoflavone, a flavonoid, is isolated from the seeds of Psoralea corylifolia. Neobavaisoflavone exhibits anti-inflammatory, anti-cancer and anti-oxidation activities. Neobavaisoflavone inhibits DNA polymerase at moderate to high concentrations. Neobavaisoflavone also inhibits platelet aggregation.

Neobavaisoflavone, a flavonoid, is isolated from the seeds of Psoralea corylifolia. Neobavaisoflavone exhibits anti-inflammatory, anti-cancer and anti-oxidation activities. Neobavaisoflavone inhibits DNA polymerase at moderate to high concentrations. Neobavaisoflavone also inhibits platelet aggregation.

Specifications

Chemical Name or Material Neobavaisoflavone
CAS 41060-15-5
Color White
Purity Grade Notes Research
Molecular Formula C20H18O4
Quantity 5 mg
Solubility Information DMSO : ≥ 31 mg/mL (96.17 mM)
SMILES O=C1C(C2=CC=C(O)C(C/C=C(C)\C)=C2)=COC3=CC(O)=CC=C13
Molecular Weight (g/mol) 322.35
Formula Weight 322.35
Percent Purity 98.74%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.