missing translation for 'onlineSavingsMsg'
Learn More

Neo Spiramycin I, TRC

Product Code. 17942845
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942845 25 mg 25mg
17900625 10 mg 10mg
17984605 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942845 Supplier Toronto Research Chemicals Supplier No. TRCN39004025MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70253-62-2
Molecular Formula C36 H62 N2 O11
Molecular Weight (g/mol) 698.88
Synonym Leucomycin V, 4A-O-de(2,6-dideoxy-3-C-methyl-α-L-ribo-hexopyranosyl)-9-O-[5-(dimethylamino)tetrahydro-6-methyl-2H-pyran-2-yl]-, [9(2R,5S,6R)]-, Neospiramycin I (6CI,7CI), 4A-O-De(2,6-dideoxy-3-C-methyl-α-L-ribo-hexopyranosyl)-9-O-[(2R,5S,6R)-5-(dimethylamino)tetrahydro-6-methyl-2H-pyran-2-yl]leucomycin V, Oxacyclohexadecane, leucomycin V deriv., Leucomycin V, 4A-O-de(2,6-dideoxy-3-C-methyl-α-L-ribo-hexopyranosyl)-9-O-[(2R,5S,6R)-5-(dimethylamino)tetrahydro-6-methyl-2H-pyran-2-yl]-, Neospiramycin I
IUPAC Name 2-[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-6-[(2S,3R,4S,5S,6R)-4-(dimethylamino)-3,5-dihydroxy-6-methyloxan-2-yl]oxy-10-[(2S,5S,6R)-5-(dimethylamino)-6-methyloxan-2-yl]oxy-4-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
SMILES CO[C@H]1[C@H](O)CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O[C@@H]2CC[C@@H]([C@@H](C)O2)N(C)C)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]3O[C@H](C)[C@@H](O)[C@@H]([C@H]3O)N(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C36H62N2O11/c1-21-19-25(17-18-39)34(49-36-33(43)31(38(7)8)32(42)24(4)47-36)35(44-9)27(40)20-29(41)45-22(2)13-11-10-12-14-28(21)48-30-16-15-26(37(5)6)23(3)46-30/h10-12,14,18,21-28,30-36,40,42-43H,13,15-17,19-20H2,1-9H3/b11-10+,14-12+/t21-,22-,23-,24-,25+,26+,27-,28+,30-,31+,32-,33-,34+,35+,36+/m1/s1
Formula Weight 698.4354
Percent Purity >95%
Chemical Name or Material Neo Spiramycin I
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.