missing translation for 'onlineSavingsMsg'
Learn More

Thomil™ Neo Quick Bactericidal and Virucidal Hydroalcoholic Disinfectant

Product Code. 17044287
Encompass
Change view
Click to view available options
Volume (Metric):
750 mL
4 L
Packaging:
Labeling: English, French, German, Italian, Portuguese, Russian, Arabic, and Spanish
Unit Size:
Pack of 2
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Packaging unitSize
17044287 750 mL Labeling: English, French, German, Italian, Portuguese, Russian, Arabic, and Spanish Pack of 6
17024287 4 L Labeling: English, French, German, Italian, Portuguese, Russian, Arabic, and Spanish Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Contains Flammable Liquids and Vapors. Keep Away from Heat, Hot Surfaces, Sparks, Open Flames and Other Sources of Ignition.
Product Code. 17044287 Supplier Thomil™ Supplier No. LSLG056EX

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Contains Flammable Liquids and Vapors. Keep Away from Heat, Hot Surfaces, Sparks, Open Flames and Other Sources of Ignition.

Neo Quick evaporates quickly with a high level of bactericidal and virucidal efficiency. Undiluted solution can be applied directly to any surface or textile.

  • Bactericidal effectiveness demonstrated against Escherichia Coli, Staphylococcus Aureus, Enterococcus Hirae, Salmonella Typhimorium, Listeria Monocytogenes and Pseudomonas Aeruginosa
  • Virucidal effectiveness demonstrated against Poliovirus, Rotavirus, Adenovirus, and Murine Norovirus
  • Product has also been tested against viruses with envelopes (Coronavirus, HIV, Influenza A H1N1, Herpesviridae, Hepatitis B, C and D, Measles, Rubella) according to UNE-EN 14476 standards
  • Solution contains a mixture of alcohols including Ethanol 65%, Isopropanol 3.1%, and Benzalkonium Chloride 0.008%.
  • Does not stain, leave veils, or have the characteristic odor of alcohol
  • Fast evaporation so that there is no need for rinsing
  • Valid for all surfaces where disinfection is necessary
  • Registered in DGSP of Spain, NHS of UK, ANSES of France, DGS and DGAV of Portugal, and PCS of Ireland

Specifications

Product Type Chemical Disinfectant
Form Liquid
pH 6.5 to 8.5
Sterility Non-sterile
Appearance Transparent
Boiling Point 84°C
Certifications/Compliance UNE-EN 13697, UNE-EN 14476
Composition Ethanol 65%, Isopropanol, 3.1%, Quaternary Ammonium 0.008%
Concentration 50 - ≤75%, 1 - ≤5%
Container Type Spray Bottle
Content And Storage 5°C to 30°C
Description NEO QUICK Bactericide & Virucide Hydroalcoholic Disinfectant
Disposable Reusable
Filtered Unfiltered
For Use With (Application) Disinfection of All Types of Surfaces
Packaging Labeling: English, French, German, Italian, Portuguese, Russian, Arabic, and Spanish
Quantity 6 Units/Box
Volume (Metric) 750 mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • neo quick
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use powder to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, propan-2-ol, quarternary ammonium compounds
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.