missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nemorubicin, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Nemorubicin (Methoxymorpholinyl doxorubicin) is a Doxorubicin derivative with potent antitumor activity. Nemorubicin is highly cytotoxic to a variety of tumor cell lines presenting a multidrug-resistant phenotype. Nemorubicin requirs an intact nucleotide excision repair (NER) system to exert its activity.
Specifications
Specifications
| Chemical Name or Material | Nemorubicin |
| CAS | 108852-90-0 |
| Color | Red |
| Purity Grade Notes | Research |
| Molecular Formula | C32H37NO13 |
| Quantity | 1 mg |
| Synonym | Methoxymorpholinyl doxorubicin FCE 23762 PNU 152243 |
| Solubility Information | DMSO : 65 mg/mL (100.99 mM; Need ultrasonic) |
| SMILES | COC1=C2C(C(C3=C(O)C(C[C@](C(CO)=O)(O)C[C@]4([H])O[C@H]5C[C@H](N6CCO[C@H](OC)C6)[C@H](O)[C@H](C)O5)=C4C(O)=C3C2=O)=O)=CC=C1 |
| Molecular Weight (g/mol) | 643.64 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction