missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Nemonapride

Product Code. 18782584
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18782584 10 mg 10mg
18742644 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782584 Supplier R&D Systems™ Supplier No. 1746/10
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent D2-like antagonist. Also 5-HT1A agonist

Highly potent dopamine D2-like receptor antagonist; selective over D1-like receptors (Ki values are 0.1 and 740 nM for D2-like and D1-like receptors respectively). Also potent 5-HT1A receptor agonist (IC50 = 34 nM) and has affinity for sigma receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75272-39-8
Molecular Formula C21H26ClN3O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 387.91
Purity >99%
Molecular Weight (g/mol) 387.908
Chemical Name or Material Nemonapride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.