missing translation for 'onlineSavingsMsg'
Learn More

Nemiralisib, MedChemExpress

Product Code. 16473646
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16473646 25 mg 25mg
16470258 5 mg 5mg
16437798 10 mg 10mg
16417636 100 mg 100mg
16411829 1 mg 1mg
16403757 50 mg 50mg
6 options
This item is not returnable. View return policy

Product Code. 16473646

Brand: Medchem Express HY19535A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nemiralisib (GSK2269557 free base) is a potent and highly selective PI3Kδ inhibitor with a pKi of 9.9.

Nemiralisib (GSK2269557 free base) is a potent and highly selective PI3Kδ inhibitor with a pKi of 9.9.

Specifications

Chemical Name or Material Nemiralisib
Physical Form Solid
CAS 1254036-71-9
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C26H28N6O
For Use With (Application) Cancer-Kinase/protease
Synonym GSK2269557 (free base)
Solubility Information DMSO : 33.33 mg/mL (75.66 mM; Need ultrasonic)
SMILES CC(N1CCN(CC2=CN=C(C3=CC(C4=CC=CC5=C4C=CN5)=CC6=C3C=NN6)O2)CC1)C
Molecular Weight (g/mol) 440.54
Formula Weight 440.54
Percent Purity 98.85%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.