missing translation for 'onlineSavingsMsg'
Learn More

Nelonemdaz, MedChemExpress

Product Code. 16476787
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476787 5 mg 5mg
16465056 100 mg 100mg
16451069 1 mg 1mg
16444088 10 mg 10mg
16420447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16476787 Supplier Medchem Express Supplier No. HY106408

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nelonemdaz (Salfaprodil free base) is an NR2B-selective and uncompetitive antagonist of N-methyl-D-aspartate (NMDA). Nelonemdaz is also a free radical scavenger. Nelonemdaz has excellent neuroprotection against NMDA- and free radical-induced cell death.

Nelonemdaz (Salfaprodil free base) is an NR2B-selective and uncompetitive antagonist of N-methyl-D-aspartate (NMDA). Nelonemdaz is also a free radical scavenger. Nelonemdaz has excellent neuroprotection against NMDA- and free radical-induced cell death.

Specifications

Chemical Name or Material Nelonemdaz
CAS 640290-67-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H8F7NO3
Quantity 5 mg
Synonym Salfaprodil free base Neu2000
Solubility Information DMSO : ≥ 112.5 mg/mL (293.57 mM)
SMILES OC(C1=C(O)C=CC(NCC2=C(C(F)=C(C(F)(F)F)C(F)=C2F)F)=C1)=O
Molecular Weight (g/mol) 383.22
Formula Weight 383.22
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.