missing translation for 'onlineSavingsMsg'
Learn More

NELL2 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30737601 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737601 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737601 Supplier Novus Biologicals Supplier No. AF6736JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

NELL2 Polyclonal antibody specifically detects NELL2 in Mouse samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NELL2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NELL2, Leu25-Leu819, Accession # NP_058023
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4753
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.