missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NELL2 Antibody (13E7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-74039CL1
This item is not returnable.
View return policy
Description
NELL2 Monoclonal antibody specifically detects NELL2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| NELL2 | |
| Monoclonal | |
| CoraFluor 1 | |
| nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2 | |
| Recombinant human NELL2 (30-258aa) purified from E. coli | |
| 0.1 mL | |
| Neuroscience | |
| 4753 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| ELISA, Western Blot | |
| 13E7 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction