missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NELL2 Antibody (055), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90179AF405
This item is not returnable.
View return policy
Description
NELL2 Monoclonal specifically detects NELL2 in Human samples. It is validated for ELISA.
Specifications
| NELL2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 55 | |
| ELISA | |
| nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human NELL2 (Q99435; Met1-Leu816). | |
| 0.1 mL | |
| Neuroscience | |
| 4753 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction