missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NELL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100288
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NELL1.
This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| NELL1 | |
| Polyclonal | |
| Unconjugated | |
| NELL1 | |
| B230343H07Rik; IDH3GL; l7R6; NELL1; NEL-like 1; NEL-like 1 (chicken); NEL-like protein 1; Nel-related protein 1; neural EGFL like 1; neural epidermal growth factor-like 1; NRP1; Protein kinase C-binding protein NELL1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 338352, 4745, 81733 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q2VWQ2, Q62919, Q92832 | |
| NELL1 | |
| A synthesized peptide derived from human NELL1(Accession Q92832), corresponding to amino acid residues A612-G662. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction