missing translation for 'onlineSavingsMsg'
Learn More

NELL1 Antibody [DyLight 680], Novus Biologicals™

Product Code. 30733606 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733606 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733606 Supplier Novus Biologicals Supplier No. AF7109Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

NELL1 Polyclonal antibody specifically detects NELL1 in Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NELL1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias FLJ45906, IDH3GL, nel (chicken)-like 1, NEL-like 1 (chicken), NEL-like protein 1, Nel-related protein 1, neural epidermal growth factor-like 1, NRP1, protein kinase C-binding protein NELL1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse NELL1, Arg17-Asn810, Accession # Q2VWQ2
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4745
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.