missing translation for 'onlineSavingsMsg'
Learn More

NELL1 Antibody (3F1), Novus Biologicals™

Product Code. 18364158 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364158 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364158 Supplier Novus Biologicals Supplier No. H00004745M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NELL1 Monoclonal antibody specifically detects NELL1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NELL1
Applications ELISA
Classification Monoclonal
Clone 3F1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006148
Gene Alias FLJ45906, IDH3GL, nel (chicken)-like 1, NEL-like 1 (chicken), NEL-like protein 1, Nel-related protein 1, neural epidermal growth factor-like 1, NRP1, protein kinase C-binding protein NELL1
Host Species Mouse
Immunogen NELL1 (NP_006148, 304 a.a. ~ 403 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CRRMSCPPLNCSPDSLPVHIAGQCCKVCRPKCIYGGKVLAEGQRILTKSCRECRGGVLVKITEMCPPLNCSEKDHILPENQCCRVCRGHNFCAEGPKCGE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4745
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.