missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nelfinavir mesylate
GREENER_CHOICE

Product Code. 18095982
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18095982

Brand: Tocris Bioscience™ 3766/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent HIV-1 protease inhibitor

Orally active human immunodeficiency virus protease inhibitor. Potently inhibits HIV-1 protease (Ki = 2 nM) in vitro. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 0.08 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159989-65-8
Molecular Formula C33H49N3O7S2

Specifications

Quantity 50 mg
Formula Weight 663.89
Purity >98%
Molecular Weight (g/mol) 663.889
Chemical Name or Material Nelfinavir mesylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.