missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nelfinavir Mesylate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Description
Nelfinavir Mesylate (AG 1343 Mesylate) is a potent and orally bioavailable HIV-1 protease inhibitor (Ki=2 nM) for HIV infection. Nelfinavir Mesylate (AG 1343 Mesylate) is a broad-spectrum, anticancer agent.
Specifications
Specifications
| Chemical Name or Material | Nelfinavir Mesylate |
| CAS | 159989-65-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C33H49N3O7S2 |
| Quantity | 5 mg |
| Synonym | AG 1343 Mesylate |
| Solubility Information | DMSO : 100 mg/mL (150.63 mM; Need ultrasonic) |
| SMILES | [H][C@]12C[C@@H](C(NC(C)(C)C)=O)N(C[C@H]([C@@H](NC(C3=C(C)C(O)=CC=C3)=O)CSC4=CC=CC=C4)O)C[C@@]1([H])CCCC2.CS(=O)(O)=O |
| Molecular Weight (g/mol) | 663.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction