missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NELFE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NELFE |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16837263
|
Proteintech
10705-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16827263
|
Proteintech
10705-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
10705-1-AP is obtained by immunizing animals with full-length RDBP of human origin.
RDBP is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The protein encoded by this gene is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6.Specifications
| NELFE | |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P18615, P19426 | |
| 27632, 294258, 7936 | |
| NELFE Fusion Protein Ag1186 | |
| Primary | |
| -20°C | |
| NELFE |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| D6S45, Negative elongation factor E, NELF E, NELFE, RD, RD RNA binding protein, RDBP, RDP, RNA binding protein RD | |
| Nelfe | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title