missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NELF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12244-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes all the isoforms.
NELF (nasal embryonic luteinizing hormone-releasing hormone factor) is a 530 amino acid transcription factor involved in the migration of LHRH neurons, outgrowth of olfactory axons and suppression of transcription elongation. NELF is found in the peripheral and central nervous system during embryonic development, and is highly expressed in adult testis, kidney and brain. Known to couple NMDA receptor signaling to the nucleus, NELF knockdown impaired GnRH neuronal migration of NLT cells in vitro and the gene encoding NELF has been linked to the development of Idiopathic hypogonadotropic hypogonadism (IHH), a disorder resulting in impaired pubertal maturation and reproductive function.Specifications
| NELF | |
| Polyclonal | |
| Unconjugated | |
| NSMF | |
| nasal embryonic LHRH factor, NELF, NSMF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117536, 26012, 56876 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6X4W1, Q99NF2, Q9EPI6 | |
| Nsmf | |
| NELF Fusion Protein Ag2888 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction