missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NELF Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542179
This item is not returnable.
View return policy
Description
Peptide sequence: GAAASRRRAL RSEAMSSVAA KVRAARAFGE YLSQSHPENR NGADHLLADA Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 79%; Rat: 100%.
NELF (nasal embryonic luteinizing hormone-releasing hormone factor) is a 530 amino acid transcription factor involved in the migration of LHRH neurons, outgrowth of olfactory axons and suppression of transcription elongation. NELF is found in the peripheral and central nervous system during embryonic development, and is highly expressed in adult testis, kidney and brain. Known to couple NMDA receptor signaling to the nucleus, NELF knockdown impaired GnRH neuronal migration of NLT cells in vitro and the gene encoding NELF has been linked to the development of Idiopathic hypogonadotropic hypogonadism (IHH), a disorder resulting in impaired pubertal maturation and reproductive function.
Specifications
| NELF | |
| Polyclonal | |
| Unconjugated | |
| NSMF | |
| HH9; Jac; Jacob; Jacob protein; Juxtasynaptic attractor of caldendrin on dendritic boutons protein; nasal embryonic LHRH factor; Nasal embryonic luteinizing hormone-releasing hormone factor; NELF; NMDA receptor synaptonuclear signaling and neuronal migration factor; NSMF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26012 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6X4W1 | |
| NSMF | |
| Synthetic peptide directed towards the N-terminal of human NELF (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction