missing translation for 'onlineSavingsMsg'
Learn More

NELF-E Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18400034 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400034 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400034 Supplier Novus Biologicals Supplier No. NBP105978AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NELF-E Polyclonal antibody specifically detects NELF-E in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NELF-E
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias D6S45, negative elongation factor E, negative elongation factor polypeptide E, NELFE, NELF-ERDmajor histocompatibility complex gene RD, nuclear protein, RD RNA binding protein, RD RNA-binding protein, RDP, RNA-binding protein RD
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human negative elongation factor polypeptide E using the numbering given in entry NP_002895.3 (GeneID 7936).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7936
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.