missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NELF Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81759JF646
This item is not returnable.
View return policy
Description
NELF Polyclonal antibody specifically detects NELF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NELF | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| nasal embryonic LHRH factorMGC125369, nasal embryonic luteinizing hormone-releasing hormone factor | |
| Rabbit polyclonal NELF antibody was raised against a 17 amino acid peptide near the center of human NELF. The immunogen is located within amino acids 230 - 280 of NELF. Amino Acid Squence: RGYAERKRRKRENDSAS | |
| 0.1 mL | |
| Neuroscience | |
| 26012 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction