missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK7 Recombinant Rabbit Monoclonal Antibody (23GB2885), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553525
This item is not returnable.
View return policy
Description
NEK7 (NIMA Related Kinase 7) is a Protein Coding gene. Diseases associated with NEK7 include Muckle-Wells Syndrome and Cinca Syndrome. Among its related pathways are Cell Cycle, Mitotic and Transport of the SLBP independent Mature mRNA. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is NEK6.
Specifications
| NEK7 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NEK7 | |
| 2810460C19Rik; AU020186; NEK7; Never in mitosis A-related kinase 7; NIMA (never in mitosis gene a)-related expressed kinase 7; NIMA (never in mitosis gene a)-related kinase 7; NIMA related kinase 7; NIMA-related expressed kinase 7; NIMA-related kinase 7; nimA-related protein kinase 7; Serine/threonine-protein kinase Nek7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140609, 360850, 59125 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2885 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D3ZBE5, Q8TDX7, Q9ES74 | |
| NEK7 | |
| A synthesized peptide derived from human NEK7 (241-302AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction