missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK6 Antibody (OTI5D7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72938JF549
This item is not returnable.
View return policy
Description
NEK6 Monoclonal specifically detects NEK6 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| NEK6 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NEK6 | |
| Full length recombinant protein of human NEK6 (NP_055212) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5D7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.11.1, Never in mitosis A-related kinase 6, NIMA (never in mitosis gene a)-related kinase 6, NimA-related protein kinase 6, Protein kinase SID6-1512, putative serine-threonine protein kinase, serine/threonine-protein kinase Nek6, SID6-1512 | |
| Mouse | |
| Affinity Purified | |
| GPCR, Protein Kinase | |
| 10783.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction