missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521458
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, H1299, U87-MG, SK-N-SH, IMR32, SK-N-AS. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NEK4 is a serine/threonine kinase involved in signal transduction and cellular communication. It is proposed to act exclusively upon threonine residues and is present in most primary carcinomas. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
| NEK4 | |
| Polyclonal | |
| Unconjugated | |
| NEK4 | |
| Nek4; Never in mitosis A-related kinase 4; NIMA (never in mitosis gene a)-related expressed kinase 4; NIMA (never in mitosis gene a)-related kinase 4; NIMA related kinase 4; NIMA-related kinase 4; nimA-related protein kinase 4; NRK2; pp12301; serine/threonine kinase 2; serine/threonine protein kinase-2; serine/threonine-protein kinase 2; Serine/threonine-protein kinase Nek4; serine/threonine-protein kinase Nek4 variant 2; serine/threonine-protein kinase NRK2; STK2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.37 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P51957 | |
| NEK4 | |
| Recombinant protein encompassing a sequence within the center region of human NEK4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction