missing translation for 'onlineSavingsMsg'
Learn More

NEK4 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18400723 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400723 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400723 Supplier Novus Biologicals Supplier No. NBP142686JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NEK4 Polyclonal antibody specifically detects NEK4 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, Never in mitosis A-related kinase 4, NIMA (never in mitosis gene a)-related kinase 4, NimA-related protein kinase 4, NRK2EC 2.7.11.1, pp12301, serine/threonine kinase 2, serine/threonine protein kinase-2, Serine/threonine-protein kinase 2, serine/threonine-protein kinase Nek4, Serine/threonine-protein kinase NRK2, STK2MGC33171
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 350 and 400 of human NIMA (never in mitosis gene a)-related kinase 4 using the numbering given in entry NP_003148.2 (GeneID 6787).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6787
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.