missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK4 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42686F
This item is not returnable.
View return policy
Description
NEK4 Polyclonal antibody specifically detects NEK4 in Human samples. It is validated for Immunohistochemistry
Specifications
| NEK4 | |
| Polyclonal | |
| Immunohistochemistry | |
| EC 2.7.11, Never in mitosis A-related kinase 4, NIMA (never in mitosis gene a)-related kinase 4, NimA-related protein kinase 4, NRK2EC 2.7.11.1, pp12301, serine/threonine kinase 2, serine/threonine protein kinase-2, Serine/threonine-protein kinase 2, serine/threonine-protein kinase Nek4, Serine/threonine-protein kinase NRK2, STK2MGC33171 | |
| The immunogen for this product maps to a region between residue 350 and 400 of human NIMA (never in mitosis gene a)-related kinase 4 using the numbering given in entry NP_003148.2 (GeneID 6787). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6787 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction