missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13649
This item is not returnable.
View return policy
Description
NEK3 Polyclonal specifically detects NEK3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NEK3 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 2.7.11, EC 2.7.11.1, glycogen synthase A kinase, HSPK 36, HSPK36, hydroxyalkyl-protein kinase, MGC29949, Never in mitosis A-related kinase 3, NIMA (never in mitosis gene a)-related kinase 3, NimA-related protein kinase 3, phosphorylase B kinase kinase, serine/threonine-protein kinase Nek3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| NEK3 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: NLILKVCQGCISPLPSHYSYELQFLVKQMFKRNPSHRPSATTLLSRGIVARLVQKCLPPEIIMEYGEEVLEEIKNSKHNTPRKKTNPSRIRIALGNEASTV | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 4752 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction