missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK2 Recombinant Rabbit Monoclonal Antibody (23GB2085), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553219
This item is not returnable.
View return policy
Description
NEK2 is involved in cell cycle regulation and is essential for faithful chromosome segregation. NEK2 is a gene that encodes for a serine/threonine-protein kinase. This protein plays a crucial role in the regulation of mitosis and is primarily located in the centrosome. During the G1 phase, the protein is undetectable, but its levels progressively increase throughout the S phase, reaching their peak in late G2 phase. The gene has several alternatively spliced transcript variants that encode different isoforms with distinct C-termini. NEK2 is associated with several diseases, including Retinitis Pigmentosa 67 and Retinitis Pigmentosa.
Specifications
| NEK2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NEK2 | |
| AA617254; C77054; HSPK 21; HsPK21; NEK 2; NEK 2A; Nek2; Nek2 kinase; NEK2A; Never in mitosis A-related kinase 2; NIMA - related expressed kinase 2; NIMA (never in mitosis gene a)-related expressed kinase 2; NIMA (never in mitosis gene a)-related kinase 2; NIMA related kinase 2; nimA-like protein kinase 1; NIMA-related expressed kinase 2; NIMA-related kinase 2; nimA-related protein kinase 2; NLK 1; NLK1; PPP1R111; protein phosphatase 1, regulatory subunit 111; RP67; Serine/threonine-protein kinase Nek2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4751 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2085 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51955 | |
| NEK2 | |
| A synthesized peptide derived from human NEK2 (21-47AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction